Product Details

SNP ID
rs76302509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136150027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCTTCCCCATAAAAATTAACC[A/G]GAAGAAGTGCATTTAAACTGAAACA
Phenotype
MIM: 614802 MIM: 604944
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSL2 PubMed Links

Gene Details

Gene
MSL2
Gene Name
male-specific lethal 2 homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145417.1 3587 UTR 3 NP_001138889.1
NM_018133.3 3587 UTR 3 NP_060603.2
XM_005247571.3 3587 UTR 3 XP_005247628.1
XM_006713684.3 3587 UTR 3 XP_006713747.1
XM_011512949.2 3587 UTR 3 XP_011511251.1
Gene
PPP2R3A
Gene Name
protein phosphatase 2 regulatory subunit B''alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190447.1 3587 Intron NP_001177376.1
NM_002718.4 3587 Intron NP_002709.2
NM_181897.2 3587 Intron NP_871626.1

View Full Product Details