Product Details

SNP ID
rs77409449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141949305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTAAATTGAAGGACGCCAAAC[A/T]TGTGGCCATTGTCTGTAGTTTCTCG
Phenotype
MIM: 602160
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TFDP2 PubMed Links

Gene Details

Gene
TFDP2
Gene Name
transcription factor Dp-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178138.1 4398 UTR 3 NP_001171609.1
NM_001178139.1 4398 UTR 3 NP_001171610.1
NM_001178140.1 4398 UTR 3 NP_001171611.1
NM_001178141.1 4398 UTR 3 NP_001171612.1
NM_001178142.1 4398 UTR 3 NP_001171613.1
NM_006286.4 4398 UTR 3 NP_006277.1
XM_005247731.2 4398 UTR 3 XP_005247788.1
XM_011513101.2 4398 UTR 3 XP_011511403.1
XM_011513102.2 4398 UTR 3 XP_011511404.1
XM_011513106.2 4398 UTR 3 XP_011511408.1
XM_011513107.1 4398 Intron XP_011511409.1
XM_011513108.2 4398 UTR 3 XP_011511410.1
XM_011513109.2 4398 UTR 3 XP_011511411.1
XM_011513111.2 4398 UTR 3 XP_011511413.1
XM_017007091.1 4398 UTR 3 XP_016862580.1
XM_017007092.1 4398 UTR 3 XP_016862581.1
XM_017007093.1 4398 UTR 3 XP_016862582.1
XM_017007094.1 4398 UTR 3 XP_016862583.1
XM_017007095.1 4398 UTR 3 XP_016862584.1
XM_017007096.1 4398 UTR 3 XP_016862585.1
XM_017007097.1 4398 UTR 3 XP_016862586.1
XM_017007098.1 4398 UTR 3 XP_016862587.1
XM_017007099.1 4398 UTR 3 XP_016862588.1
XM_017007100.1 4398 Intron XP_016862589.1
XM_017007101.1 4398 UTR 3 XP_016862590.1
XM_017007102.1 4398 UTR 3 XP_016862591.1
XM_017007103.1 4398 Intron XP_016862592.1
XM_017007104.1 4398 Intron XP_016862593.1
XM_017007105.1 4398 Intron XP_016862594.1

View Full Product Details