Product Details

SNP ID
rs79709647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169972005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTCTATGTGTTCTACATTTGAA[A/G]TGTTTCAGATATCTAAATTTCTTTC
Phenotype
MIM: 602173
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100128164 PubMed Links

Gene Details

Gene
LOC100128164
Gene Name
four and a half LIM domains 1 pseudogene
There are no transcripts associated with this gene.

Gene
SEC62
Gene Name
SEC62 homolog, preprotein translocation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003262.3 Intron NP_003253.1
XM_011513114.2 Intron XP_011511416.1

View Full Product Details