Product Details

SNP ID
rs75079502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179333118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATTCTAGACCACACTGCCTTTT[C/T]TACAGGTTGGGGGAACTAATTTATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF639 PubMed Links

Gene Details

Gene
ZNF639
Gene Name
zinc finger protein 639
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303425.1 1044 Missense Mutation TCT,TTT S100F NP_001290354.1
NM_001303426.1 1044 Missense Mutation TCT,TTT S100F NP_001290355.1
NM_016331.2 1044 Missense Mutation TCT,TTT S100F NP_057415.1
XM_017006550.1 1044 Missense Mutation TCT,TTT S100F XP_016862039.1
XM_017006551.1 1044 Missense Mutation TCT,TTT S100F XP_016862040.1
XM_017006552.1 1044 Missense Mutation TCT,TTT S100F XP_016862041.1
XM_017006553.1 1044 Missense Mutation TCT,TTT S100F XP_016862042.1

View Full Product Details