Product Details

SNP ID
rs74656805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185078347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTGTTCAGGCTACTAAGAGAA[C/T]ACTGCTTCAACCTTAATCGAAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf70 PubMed Links

Gene Details

Gene
C3orf70
Gene Name
chromosome 3 open reading frame 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025266.1 5604 UTR 3 NP_001020437.1

View Full Product Details