Product Details

SNP ID
rs80221956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4267723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTAGAAGTTTGGACCAGAATA[C/T]ATTTTATTTTTCTCATAAAAGTTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYAR PubMed Links

Gene Details

Gene
LYAR
Gene Name
Ly1 antibody reactive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145725.1 1480 UTR 3 NP_001139197.1
NM_017816.2 1480 UTR 3 NP_060286.1
XM_011513505.1 1480 UTR 3 XP_011511807.1
XM_011513506.2 1480 UTR 3 XP_011511808.1

View Full Product Details