Product Details

SNP ID
rs75361409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40424483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAACCTTTTTAATAACATTGAAA[C/T]AAAATGGTTTCATTTTGCATTAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM47 PubMed Links

Gene Details

Gene
RBM47
Gene Name
RNA binding motif protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098634.1 4034 UTR 3 NP_001092104.1
NM_019027.3 4034 UTR 3 NP_061900.2
XM_005248103.3 4034 UTR 3 XP_005248160.1
XM_005248107.3 4034 UTR 3 XP_005248164.1
XM_005248108.3 4034 UTR 3 XP_005248165.1
XM_005248109.4 4034 UTR 3 XP_005248166.1
XM_011513704.2 4034 UTR 3 XP_011512006.1
XM_011513707.2 4034 UTR 3 XP_011512009.1
XM_011513708.2 4034 UTR 3 XP_011512010.1
XM_017008304.1 4034 UTR 3 XP_016863793.1
XM_017008305.1 4034 UTR 3 XP_016863794.1
XM_017008306.1 4034 UTR 3 XP_016863795.1
XM_017008307.1 4034 UTR 3 XP_016863796.1
XM_017008308.1 4034 UTR 3 XP_016863797.1
XM_017008309.1 4034 UTR 3 XP_016863798.1
XM_017008310.1 4034 UTR 3 XP_016863799.1

View Full Product Details