Product Details

SNP ID
rs78454293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:192352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGACCCCCTCCTCGCCTCCATC[A/G]CCCGGGAGCTCAGCAAGATGGCGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC14B PubMed Links

Gene Details

Gene
LRRC14B
Gene Name
leucine rich repeat containing 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080478.1 842 Missense Mutation ACC,GCC T272A NP_001073947.1
Gene
PLEKHG4B
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
There are no transcripts associated with this gene.

View Full Product Details