Product Details
- SNP ID
-
rs78964548
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:35628504 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATTTTCCAGTGGCTATCTACTT[G/T]GAGAAGTTCTACACAAGTTTGAACT
- Phenotype
-
MIM: 610172
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SPEF2
PubMed Links
Gene Details
- Gene
- SPEF2
- Gene Name
- sperm flagellar 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024867.3 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
NP_079143.3 |
NM_144722.3 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
NP_653323.1 |
XM_005248376.3 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_005248433.1 |
XM_005248377.3 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_005248434.1 |
XM_005248378.3 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_005248435.1 |
XM_011514135.2 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512437.1 |
XM_011514136.2 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512438.1 |
XM_011514137.2 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512439.1 |
XM_011514138.2 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512440.1 |
XM_011514139.2 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512441.1 |
XM_011514140.2 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512442.1 |
XM_011514141.2 |
323 |
UTR 5 |
|
|
XP_011512443.1 |
XM_011514142.1 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_011512444.1 |
XM_017009880.1 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_016865369.1 |
XM_017009881.1 |
323 |
UTR 5 |
|
|
XP_016865370.1 |
XM_017009882.1 |
323 |
Nonsense Mutation |
GGA,TGA |
G35* |
XP_016865371.1 |
View Full Product Details