Product Details
- SNP ID
-
rs79063860
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:38940069 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACCACAAAGCTCTGAGTATCTCC[A/C]CAAAGTAAGTGATATAGGCAGATAT
- Phenotype
-
MIM: 601743
MIM: 609022
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OSMR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149159011] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OSMR
- Gene Name
- oncostatin M receptor
- Gene
- RICTOR
- Gene Name
- RPTOR independent companion of MTOR complex 2
View Full Product Details