Product Details

SNP ID
rs79164838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77629307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATGACTGCTTTAGTCTACTCC[A/T]GCCACCCAATCCACTGTCACGTTCC
Phenotype
MIM: 604529
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OTP PubMed Links

Gene Details

Gene
OTP
Gene Name
orthopedia homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032109.2 2083 UTR 3 NP_115485.1
Gene
WDR41
Gene Name
WD repeat domain 41
There are no transcripts associated with this gene.

View Full Product Details