Product Details

SNP ID
rs79309522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:86619134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCTCAAGACCATTCATTGCCT[A/C]GTGCTTAGTAAAGCCAGATTATTTC
Phenotype
MIM: 603774
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COX7C PubMed Links

Gene Details

Gene
COX7C
Gene Name
cytochrome c oxidase subunit 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001867.2 Intron NP_001858.1
Gene
MIR3607
Gene Name
microRNA 3607
There are no transcripts associated with this gene.

View Full Product Details