Product Details

SNP ID
rs79395430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:86620271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCGTTCCAACTTGGGGGCAAAA[C/T]TACCAGTGGTGCTGAGGTTGAGAAG
Phenotype
MIM: 603774
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX7C PubMed Links

Gene Details

Gene
COX7C
Gene Name
cytochrome c oxidase subunit 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001867.2 Intron NP_001858.1
Gene
MIR3607
Gene Name
microRNA 3607
There are no transcripts associated with this gene.

View Full Product Details