Product Details

SNP ID
rs75792646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82363311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGCGGGAAGGCATTAATTACA[C/T]CTAAATCTGAAGTTTGCGGCCTCAA
Phenotype
MIM: 190920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPBG PubMed Links

Gene Details

Gene
TPBG
Gene Name
trophoblast glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166392.1 106 Intron NP_001159864.1
NM_006670.4 106 UTR 5 NP_006661.1
XM_011536097.2 106 Intron XP_011534399.1

View Full Product Details