Product Details

SNP ID
rs74444948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17602595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTTTTTTAATTTACAGGCAGA[G/T]AATATGTTATTCCATCCTTGGCCCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM8A1 PubMed Links

Gene Details

Gene
FAM8A1
Gene Name
family with sequence similarity 8 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016255.2 841 Nonsense Mutation GAA,TAA E240* NP_057339.1

View Full Product Details