Product Details
- SNP ID
-
rs77679854
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:28162571 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTTTTTCTTTGAGTAAAATGTTT[C/G]TTTTACAGAAGTGTTCCTGAAAACT
- Phenotype
-
MIM: 602240
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZKSCAN8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72087025] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZKSCAN8
- Gene Name
- zinc finger with KRAB and SCAN domains 8
There are no transcripts associated with this gene.
- Gene
- ZNF192P1
- Gene Name
- zinc finger protein 192 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details