Product Details

SNP ID
rs77742720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28259697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCAGAAGAACGGCAGTCAGCGG[A/G]AGACTACGAGAAGGAAGAGAGCCAT
Phenotype
MIM: 611643 MIM: 616474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKAPL PubMed Links

Gene Details

Gene
NKAPL
Gene Name
NFKB activating protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007531.2 401 Missense Mutation GAA,GGA E109G NP_001007532.1
Gene
ZKSCAN4
Gene Name
zinc finger with KRAB and SCAN domains 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304506.1 401 Intron NP_001291435.1
NM_019110.4 401 Intron NP_061983.2
XM_005249095.3 401 Intron XP_005249152.1
XM_005249097.3 401 Intron XP_005249154.1
XM_005249098.3 401 Intron XP_005249155.1
XM_005249100.4 401 Intron XP_005249157.2
XM_006715095.3 401 Intron XP_006715158.1
XM_011514588.2 401 Intron XP_011512890.1
XM_011514589.2 401 Intron XP_011512891.1
XM_017010844.1 401 Intron XP_016866333.1
Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
There are no transcripts associated with this gene.

View Full Product Details