Product Details

SNP ID
rs78184239
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28378911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGTAATAGAGTTAAAAAAAAC[G/T]GTGTGATTTGCAGATACTGGAAAAG
Phenotype
MIM: 612791 MIM: 603978
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZKSCAN3 PubMed Links

Gene Details

Gene
ZKSCAN3
Gene Name
zinc finger with KRAB and SCAN domains 3
There are no transcripts associated with this gene.

Gene
ZSCAN12
Gene Name
zinc finger and SCAN domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163391.1 7219 Intron NP_001156863.1
XM_011515014.2 7219 UTR 3 XP_011513316.1
XM_011515015.2 7219 UTR 3 XP_011513317.1
XM_011515017.2 7219 UTR 3 XP_011513319.1
XM_017011528.1 7219 UTR 3 XP_016867017.1

View Full Product Details