Product Details

SNP ID
rs78855526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32196984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAATCAGTTCTTCAACCAGGTCT[C/T]CCACCGCCAGCCTGGCAGCCAGCAT
Phenotype
MIM: 164951 MIM: 176311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPSM3 PubMed Links

Gene Details

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
There are no transcripts associated with this gene.

Gene
NOTCH4
Gene Name
notch 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004557.3 5280 Missense Mutation GAA,GGA E1714G NP_004548.3
Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

View Full Product Details