Product Details

SNP ID
rs77298778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39796500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAACTGCACAGTCAAGATGCTGT[A/G]GGGGGAGGAAGTAAGTGCCTGTTCT
Phenotype
MIM: 606627
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAAM2 PubMed Links

Gene Details

Gene
DAAM2
Gene Name
dishevelled associated activator of morphogenesis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201427.1 Intron NP_001188356.1
NM_015345.3 Intron NP_056160.2
XM_006715039.3 Intron XP_006715102.3
XM_006715040.3 Intron XP_006715103.3
XM_006715042.2 Intron XP_006715105.1
XM_006715043.2 Intron XP_006715106.1
XM_006715045.3 Intron XP_006715108.1
XM_006715046.3 Intron XP_006715109.1
XM_017010630.1 Intron XP_016866119.1

View Full Product Details