Product Details

SNP ID
rs78305894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41798807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCATGACCACTGCGGAGAGATCA[C/T]AGGCAAAGGTCTCTTTGTCAAGAGA
Phenotype
MIM: 616168
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOMM6 PubMed Links

Gene Details

Gene
TOMM6
Gene Name
translocase of outer mitochondrial membrane 6
There are no transcripts associated with this gene.

Gene
USP49
Gene Name
ubiquitin specific peptidase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286554.1 1895 Missense Mutation TAT,TGT Y598C NP_001273483.1
NM_018561.4 1895 Missense Mutation TAT,TGT Y598C NP_061031.2

View Full Product Details