Product Details

SNP ID
rs75651354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42747736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTTACTCTTGTGCATAGTCAT[C/T]GTTTTCACTAGTCTTGTTTTATGGG
Phenotype
MIM: 602971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLTSCR1L PubMed Links

Gene Details

Gene
GLTSCR1L
Gene Name
GLTSCR1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318819.1 Intron NP_001305748.1
NM_015349.2 Intron NP_056164.1
Gene
TBCC
Gene Name
tubulin folding cofactor C
There are no transcripts associated with this gene.

View Full Product Details