Product Details

SNP ID
rs75255431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52742446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACAGCCACATTCAGTGCTTTT[C/T]AGCACATTCACAGAACTCTGCAAAA
Phenotype
MIM: 138360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
There are no transcripts associated with this gene.

Gene
GSTA7P
Gene Name
glutathione S-transferase alpha 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details