Product Details

SNP ID
rs78014243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29151865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGAAATTGTATGTATAGCCATCA[A/G]CACAATGTCTGGCCCATAGCAAGCA
Phenotype
MIM: 602857 MIM: 609780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHN2 PubMed Links

Gene Details

Gene
CHN2
Gene Name
chimerin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039936.2 Intron NP_001035025.1
NM_001293069.1 Intron NP_001279998.1
NM_001293070.1 Intron NP_001279999.1
NM_001293071.1 Intron NP_001280000.1
NM_001293072.1 Intron NP_001280001.1
NM_001293073.1 Intron NP_001280002.1
NM_001293075.1 Intron NP_001280004.1
NM_001293076.1 Intron NP_001280005.1
NM_001293077.1 Intron NP_001280006.1
NM_001293078.1 Intron NP_001280007.1
NM_001293079.1 Intron NP_001280008.1
NM_001293080.1 Intron NP_001280009.1
NM_001293081.1 Intron NP_001280010.1
NM_004067.3 Intron NP_004058.1
XM_011515105.2 Intron XP_011513407.1
XM_011515106.2 Intron XP_011513408.1
XM_011515107.2 Intron XP_011513409.1
XM_017011721.1 Intron XP_016867210.1
XM_017011722.1 Intron XP_016867211.1
Gene
CPVL
Gene Name
carboxypeptidase, vitellogenic like
There are no transcripts associated with this gene.

View Full Product Details