Product Details

SNP ID
rs79436512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:32485382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGCAAAGGTTTTCCAATCCAC[A/G]TGGACCAAATACAAAAACCATAATG
Phenotype
MIM: 607285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSM5 PubMed Links

Gene Details

Gene
LSM5
Gene Name
LSM5 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130710.1 2201 UTR 3 NP_001124182.1
NM_001139499.1 2201 UTR 3 NP_001132971.1
NM_012322.2 2201 UTR 3 NP_036454.1

View Full Product Details