Product Details

SNP ID
rs77356059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44051169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGACTGCCTCCACTCATACGGAG[C/T]CCTGTAGATGAAATACCTGATCAGC
Phenotype
MIM: 610106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DBNL PubMed Links

Gene Details

Gene
DBNL
Gene Name
drebrin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014436.2 Intron NP_001014436.1
NM_001122956.1 Intron NP_001116428.1
NM_001284313.1 Intron NP_001271242.1
NM_001284315.1 Intron NP_001271244.1
NM_014063.6 Intron NP_054782.2
XM_011515325.1 Intron XP_011513627.1
Gene
LINC00957
Gene Name
long intergenic non-protein coding RNA 957
There are no transcripts associated with this gene.

Gene
MIR6837
Gene Name
microRNA 6837
There are no transcripts associated with this gene.

View Full Product Details