Product Details

SNP ID
rs76861115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6618954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTCTATGAAAAGCACGCTCAC[A/G]GATCTGTTAGCCTAACCAAAGGTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF853 PubMed Links

Gene Details

Gene
ZNF853
Gene Name
zinc finger protein 853
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017560.1 Intron NP_060030.1
XM_011515438.2 Intron XP_011513740.1
XM_011515439.2 Intron XP_011513741.1
XM_017012371.1 Intron XP_016867860.1

View Full Product Details