Product Details

SNP ID
rs78364029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:86880388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGATCACCAATGTGACAGGTA[C/T]GAGGACAGACCCTAAAATATGGTCC
Phenotype
MIM: 614048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1324L PubMed Links

Gene Details

Gene
KIAA1324L
Gene Name
KIAA1324 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142749.2 3358 UTR 3 NP_001136221.1
NM_001291990.1 3358 UTR 3 NP_001278919.1
NM_001291991.1 3358 UTR 3 NP_001278920.1
NM_152748.3 3358 UTR 3 NP_689961.3
XM_006715894.3 3358 UTR 3 XP_006715957.1
XM_011515918.2 3358 UTR 3 XP_011514220.1
XM_011515921.1 3358 UTR 3 XP_011514223.1
XM_017011843.1 3358 UTR 3 XP_016867332.1
XM_017011844.1 3358 UTR 3 XP_016867333.1

View Full Product Details