Product Details

SNP ID
rs76665876
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99892234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAACCAAAAAAATGGCCGGAGGC[A/G]TGAGACAGAAGAACAAGAAAAGGTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TRIM4 PubMed Links

Gene Details

Gene
TRIM4
Gene Name
tripartite motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033017.3 1553 Missense Mutation CGC,TGC R478C NP_148977.2
NM_033091.2 1553 Missense Mutation CGC,TGC R452C NP_149082.1

View Full Product Details