Product Details

SNP ID
rs80338354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100436374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGACTCCCCACAACCCATCATCA[G/T]CTTTGGAGGTGCCTTGAGGGTGCGG
Phenotype
MIM: 611478
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MEPCE PubMed Links

Gene Details

Gene
MEPCE
Gene Name
methylphosphate capping enzyme
There are no transcripts associated with this gene.

Gene
PPP1R35
Gene Name
protein phosphatase 1 regulatory subunit 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145030.2 98 Missense Mutation NP_659467.1
XM_011515914.1 98 Intron XP_011514216.1
Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
There are no transcripts associated with this gene.

View Full Product Details