Product Details

SNP ID
rs76320540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128310296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGAAAGCATTTCTCTCTGCTCT[C/T]GCTTAGACAGTTCCTGCAGTGTGTA
Phenotype
MIM: 612074
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGC27345 PubMed Links

Gene Details

Gene
MGC27345
Gene Name
uncharacterized protein MGC27345
There are no transcripts associated with this gene.

Gene
RBM28
Gene Name
RNA binding motif protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166135.1 2671 Intron NP_001159607.1
NM_018077.2 2671 Intron NP_060547.2
XM_011516370.2 2671 UTR 3 XP_011514672.1
XM_017012389.1 2671 UTR 3 XP_016867878.1
XM_017012390.1 2671 UTR 3 XP_016867879.1

View Full Product Details