Product Details

SNP ID
rs78305161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130486718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACCCTCGCCGAGGCTTGACCCC[C/T]GCGACAGAGTTTAGTCATTCCGTTA
Phenotype
MIM: 601029 MIM: 607794 MIM: 611768
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEST PubMed Links

Gene Details

Gene
MEST
Gene Name
mesoderm specific transcript
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253900.1 Intron NP_001240829.1
NM_001253901.1 Intron NP_001240830.1
NM_001253902.1 Intron NP_001240831.1
NM_002402.3 Intron NP_002393.2
NM_177524.2 Intron NP_803490.1
NM_177525.2 Intron NP_803491.1
XM_011516222.1 Intron XP_011514524.1
XM_017012218.1 Intron XP_016867707.1
Gene
MESTIT1
Gene Name
MEST intronic transcript 1, antisense RNA
There are no transcripts associated with this gene.

Gene
MIR335
Gene Name
microRNA 335
There are no transcripts associated with this gene.

View Full Product Details