Product Details

SNP ID
rs76010286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123181425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACACAATCCTGATTTCTTGGT[A/G]TCGTCCCTGAGCAGATAGCGTCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83A PubMed Links

Gene Details

Gene
FAM83A
Gene Name
family with sequence similarity 83 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288587.2 Intron NP_001275516.1
NM_001321630.1 Intron NP_001308559.1
NM_032899.6 Intron NP_116288.2
NM_207006.2 Intron NP_996889.1
XM_005251087.2 Intron XP_005251144.1
XM_011517338.1 Intron XP_011515640.1

View Full Product Details