Product Details

SNP ID
rs76268228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142703109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTAGAAATATTTCCACGTTTTT[C/T]CATGGTTGCGAAGCAGTGCTCCGCT
Phenotype
MIM: 615093
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
LY6K
Gene Name
lymphocyte antigen 6 complex, locus K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160354.1 900 UTR 3 NP_001153826.1
NM_001160355.1 900 Intron NP_001153827.1
NM_017527.3 900 Missense Mutation TCC,TTC S79F NP_059997.3
XM_005250989.4 900 Intron XP_005251046.1
XM_005250990.4 900 UTR 3 XP_005251047.1

View Full Product Details