Product Details

SNP ID
rs74523522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:65718469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTCTCGCATGTGCAGGTCAA[C/G]TGTACAGTAGTGAAAACAACATGCA
Phenotype
MIM: 171885
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MTFR1 PubMed Links

Gene Details

Gene
MTFR1
Gene Name
mitochondrial fission regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145838.1 2345 Intron NP_001139310.1
NM_001145839.1 2345 Intron NP_001139311.1
NM_014637.3 2345 Intron NP_055452.3
XM_006716484.2 2345 Intron XP_006716547.2
XM_011517626.1 2345 Intron XP_011515928.1
XM_011517627.2 2345 Intron XP_011515929.1
XM_011517628.2 2345 Intron XP_011515930.1
Gene
PDE7A
Gene Name
phosphodiesterase 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242318.2 2345 UTR 3 NP_001229247.1
NM_002603.3 2345 UTR 3 NP_002594.1
XM_011517540.2 2345 UTR 3 XP_011515842.1
XM_017013538.1 2345 UTR 3 XP_016869027.1

View Full Product Details