Product Details

SNP ID
rs77388689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80971229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCTTTTGAAACAATCAATGAAA[A/G]TGCTACTAAGTGTCATGAATTATTT
Phenotype
MIM: 605767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAG1 PubMed Links

Gene Details

Gene
PAG1
Gene Name
phosphoprotein membrane anchor with glycosphingolipid microdomains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018440.3 7278 UTR 3 NP_060910.3
XM_017013640.1 7278 UTR 3 XP_016869129.1
XM_017013641.1 7278 UTR 3 XP_016869130.1
XM_017013642.1 7278 UTR 3 XP_016869131.1
XM_017013643.1 7278 UTR 3 XP_016869132.1

View Full Product Details