Product Details

SNP ID
rs78267433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35813450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCCCTGTTCTTCCCACCTGCT[G/T]GTCTTCTTCTTCAGCCTGGCTAATC
Phenotype
MIM: 609997 MIM: 108961 MIM: 605731
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM221B PubMed Links

Gene Details

Gene
FAM221B
Gene Name
family with sequence similarity 221 member B
There are no transcripts associated with this gene.

Gene
HINT2
Gene Name
histidine triad nucleotide binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032593.2 388 Missense Mutation AAG,CAG K108Q NP_115982.1
Gene
NPR2
Gene Name
natriuretic peptide receptor 2
There are no transcripts associated with this gene.

Gene
SPAG8
Gene Name
sperm associated antigen 8
There are no transcripts associated with this gene.

View Full Product Details