Product Details

SNP ID
rs79928821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86027419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGTACAGCACGTGGACAGGAC[A/G]ACGGAACCCAGAGTTCTCTGTCTCT
Phenotype
MIM: 606804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLM1 PubMed Links

Gene Details

Gene
GOLM1
Gene Name
golgi membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016548.3 1802 UTR 3 NP_057632.2
NM_177937.2 1802 UTR 3 NP_808800.1
Gene
NAA35
Gene Name
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
There are no transcripts associated with this gene.

View Full Product Details