Product Details

SNP ID
rs75111780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92845553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTAATTTGTCCTATTTTTCAAA[C/T]AGGAGTTGGTAACAACAACATCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD19P PubMed Links

Gene Details

Gene
ANKRD19P
Gene Name
ankyrin repeat domain 19, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF484
Gene Name
zinc finger protein 484
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007101.2 3368 UTR 3 NP_001007102.1
NM_001261458.1 3368 UTR 3 NP_001248387.1
NM_001261459.1 3368 UTR 3 NP_001248388.1
NM_001261460.1 3368 UTR 3 NP_001248389.1
NM_031486.2 3368 UTR 3 NP_113674.1
XM_006717299.3 3368 UTR 3 XP_006717362.1
XM_011519073.2 3368 UTR 3 XP_011517375.1
XM_011519074.2 3368 UTR 3 XP_011517376.1
XM_011519075.2 3368 UTR 3 XP_011517377.1
XM_017015178.1 3368 UTR 3 XP_016870667.1
XM_017015179.1 3368 UTR 3 XP_016870668.1
XM_017015180.1 3368 UTR 3 XP_016870669.1
XM_017015181.1 3368 UTR 3 XP_016870670.1

View Full Product Details