Product Details

SNP ID
rs75779550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93447234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCTTTTTTTCTATGCAAGTAT[C/G]TATTTGTCTATATGTCTGTCTACCT
Phenotype
MIM: 612265
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM120A PubMed Links

Gene Details

Gene
FAM120A
Gene Name
family with sequence similarity 120A
There are no transcripts associated with this gene.

Gene
FAM120AOS
Gene Name
family with sequence similarity 120A opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322224.1 2031 UTR 3 NP_001309153.1
NM_198841.2 2031 UTR 3 NP_942138.2

View Full Product Details