Product Details

SNP ID
rs77345671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93452234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGCTCGAGAAGGCCCGGCTGCAC[A/G]AGTGGGTCAAGCGGCAGGGCAACGA
Phenotype
MIM: 612265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM120A PubMed Links

Gene Details

Gene
FAM120A
Gene Name
family with sequence similarity 120A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286722.1 1359 Missense Mutation AAG,GAG K107E NP_001273651.1
NM_001286723.1 1359 Missense Mutation AAG,GAG K107E NP_001273652.1
NM_001286724.1 1359 Missense Mutation AAG,GAG K107E NP_001273653.1
NM_014612.4 1359 Missense Mutation AAG,GAG K107E NP_055427.2
XM_005251842.4 1359 Missense Mutation AAG,GAG K107E XP_005251899.1
XM_011518412.2 1359 Missense Mutation AAG,GAG K107E XP_011516714.1
XM_011518413.2 1359 Missense Mutation AAG,GAG K107E XP_011516715.1
XM_011518414.2 1359 Missense Mutation AAG,GAG K107E XP_011516716.1
XM_011518416.2 1359 Missense Mutation AAG,GAG K107E XP_011516718.1
Gene
FAM120AOS
Gene Name
family with sequence similarity 120A opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322224.1 1359 Intron NP_001309153.1
NM_198841.2 1359 Missense Mutation TCG,TTG S159L NP_942138.2

View Full Product Details