Product Details
- SNP ID
-
rs75107690
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:94190148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATACCATACGTTGTCACTTACAA[A/G]CTTAGTTCTCACTTATAAGTGACAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIRLET7DHG
PubMed Links
Gene Details
- Gene
- MIRLET7DHG
- Gene Name
- MIRLET7D host gene
There are no transcripts associated with this gene.
View Full Product Details