Product Details

SNP ID
rs77924894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72124040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAACACTTGAGAGTATTGTAC[C/T]ATAGTTTTGCAGAGAGGAAAACAAG
Phenotype
MIM: 139260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDA PubMed Links

Gene Details

Gene
GDA
Gene Name
guanine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242505.2 Intron NP_001229434.1
NM_001242506.2 Intron NP_001229435.1
NM_001242507.2 Intron NP_001229436.1
NM_004293.4 Intron NP_004284.1
XM_005252317.1 Intron XP_005252374.1
XM_011519213.1 Intron XP_011517515.1
XM_011519214.1 Intron XP_011517516.1
XM_011519215.1 Intron XP_011517517.1
XM_011519216.1 Intron XP_011517518.1
XM_011519217.1 Intron XP_011517519.1
XM_011519218.2 Intron XP_011517520.1
XM_011519219.1 Intron XP_011517521.1
XM_011519220.1 Intron XP_011517522.1
XM_011519221.1 Intron XP_011517523.1
XM_011519222.1 Intron XP_011517524.1
XM_011519223.2 Intron XP_011517525.1
XM_017015333.1 Intron XP_016870822.1
XM_017015334.1 Intron XP_016870823.1
XM_017015335.1 Intron XP_016870824.1
XM_017015336.1 Intron XP_016870825.1
XM_017015337.1 Intron XP_016870826.1
XM_017015338.1 Intron XP_016870827.1
XM_017015339.1 Intron XP_016870828.1
XM_017015340.1 Intron XP_016870829.1

View Full Product Details