Product Details

SNP ID
rs79206720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122163630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACGTATGGAGTAGAGCTGGCAG[C/T]TCAGTTTGGCAGGAGAGTGGGTTGT
Phenotype
MIM: 603359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORN5 PubMed Links

Gene Details

Gene
MORN5
Gene Name
MORN repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286828.1 Intron NP_001273757.1
NM_198469.3 Intron NP_940871.2
XM_005251877.4 Intron XP_005251934.1
XM_005251878.3 Intron XP_005251935.1
XM_006717034.3 Intron XP_006717097.1
XM_011518457.2 Intron XP_011516759.1
XM_011518458.1 Intron XP_011516760.1
XM_017014578.1 Intron XP_016870067.1
Gene
NDUFA8
Gene Name
NADH:ubiquinone oxidoreductase subunit A8
There are no transcripts associated with this gene.

View Full Product Details