Product Details

SNP ID
rs78597609
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124458659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCAGCCATCATCTCCTCTGAA[C/G]TGTGGGACGTCACTGGAGAGGTCAA
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ADGRD2 PubMed Links

Gene Details

Gene
ADGRD2
Gene Name
adhesion G protein-coupled receptor D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006717348.3 1786 Missense Mutation CTG,GTG L588V XP_006717411.2

View Full Product Details