Product Details

SNP ID
rs79199260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130184365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTGGTTAGTTCTGGGACCTTGG[C/T]CTGGCCTTTCCCACCGAGCCCTCAG
Phenotype
MIM: 603315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCS1 PubMed Links

Gene Details

Gene
NCS1
Gene Name
neuronal calcium sensor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128826.1 Intron NP_001122298.1
NM_014286.3 Intron NP_055101.2

View Full Product Details