Product Details

SNP ID
rs78641202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132210052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAACCCTATCTAGACATACAAAA[C/T]GTACAGCAAAACCCAGTGTTACCAT
Phenotype
MIM: 608407
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYSL4 PubMed Links

Gene Details

Gene
DPYSL4
Gene Name
dihydropyrimidinase like 4
There are no transcripts associated with this gene.

Gene
STK32C
Gene Name
serine/threonine kinase 32C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318878.1 Intron NP_001305807.1
NM_001318879.1 Intron NP_001305808.1
NM_001318880.1 Intron NP_001305809.1
NM_001318881.1 Intron NP_001305810.1
NM_001318882.1 Intron NP_001305811.1
NM_001318883.1 Intron NP_001305812.1
NM_173575.3 Intron NP_775846.2
XM_011539688.1 Intron XP_011537990.1
XM_011539690.2 Intron XP_011537992.1
XM_011539693.2 Intron XP_011537995.1
XM_011539694.1 Intron XP_011537996.1
XM_011539695.1 Intron XP_011537997.1
XM_011539696.1 Intron XP_011537998.1
XM_017016100.1 Intron XP_016871589.1
XM_017016101.1 Intron XP_016871590.1

View Full Product Details