Product Details

SNP ID
rs77411787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1557802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGAAGGTGGTACCTGTAGGC[A/G]TCGTCGGAGGACATGCCCATGGTCT
Phenotype
MIM: 602038
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP8 PubMed Links

Gene Details

Gene
DUSP8
Gene Name
dual specificity phosphatase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004420.2 1968 Silent Mutation GAC,GAT D271D NP_004411.2
XM_011519932.2 1968 Silent Mutation GAC,GAT D271D XP_011518234.1
XM_011519933.2 1968 Silent Mutation GAC,GAT D271D XP_011518235.1
Gene
LOC101927562
Gene Name
uncharacterized LOC101927562
There are no transcripts associated with this gene.

View Full Product Details