Product Details

SNP ID
rs76952436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16742784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGTGAATTTAACAATAGCAAAC[C/T]GAAATTTATATTCTTTGTTTTCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf58 PubMed Links

Gene Details

Gene
C11orf58
Gene Name
chromosome 11 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014267.5 1617 Intron NP_055082.1
XM_017017143.1 1617 UTR 5 XP_016872632.1

View Full Product Details