Product Details
- SNP ID
-
rs79861273
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:27519467 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTGGTGACAATGGTGCAATGGTA[A/G]CATAGAAGTAATAATTTGGATGCTT
- Phenotype
-
MIM: 611468
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BDNF-AS
PubMed Links
Gene Details
- Gene
- BDNF-AS
- Gene Name
- BDNF antisense RNA
There are no transcripts associated with this gene.
- Gene
- MIR8087
- Gene Name
- microRNA 8087
There are no transcripts associated with this gene.
View Full Product Details